Assay Details
| Gene Symbol: dlat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269021.4 | NP_001255950.1 | 9 - 10 | 1643 | 82 | |
| Gene Symbol: C30H6.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383265.1 | 8 - 9 | 1562 | 82 | ||
More Information
Set Membership: |
|

dlat-2
Genomic Map