Assay Details
| Gene Symbol: egl-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026176.3 | NP_001021347.1 | 16 - 17 | 3715 | 68 | |
| NM_001313085.3 | NP_001300014.1 | 15 | 3776 | 68 | ||
| NM_001313087.3 | NP_001300016.1 | 16 - 17 | 3640 | 68 | ||
| NM_001313089.4 | NP_001300018.1 | 15 - 16 | 3404 | 68 | ||
| NM_001313091.3 | NP_001300020.1 | 15 - 16 | 3466 | 68 | ||
| NM_001313093.3 | NP_001300022.1 | 14 - 15 | 3223 | 68 | ||
| NM_001313095.4 | NP_001300024.1 | 15 | 3827 | 68 | ||
| NM_001313097.3 | NP_001300026.1 | 13 | 3449 | 68 | ||
| NM_001313099.3 | NP_001300028.1 | 13 | 3374 | 68 | ||
| NM_001313101.3 | NP_001300030.1 | 12 | 3131 | 68 | ||
| NM_001313103.4 | NP_001300032.1 | 14 - 15 | 3376 | 68 | ||
| NM_001313105.3 | NP_001300034.1 | 11 | 2951 | 68 | ||
| GenBank mRNA | AF179426.1 | - | 16 - 17 | 3715 | 68 | |
More Information
Set Membership: |
|

egl-8
Genomic Map