Assay Details
| Gene Symbol: egl-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026175.4 | NP_001021346.1 | 9 - 10 | 2093 | 63 | |
| NM_001026176.3 | NP_001021347.1 | 9 - 10 | 2093 | 63 | ||
| NM_001313085.3 | NP_001300014.1 | 9 | 1911 | 63 | ||
| NM_001313095.4 | NP_001300024.1 | 8 | 1962 | 63 | ||
| NM_001313097.3 | NP_001300026.1 | 8 | 1827 | 63 | ||
| NM_001380576.2 | NP_001368552.1 | 9 - 10 | 1911 | 63 | ||
| NM_001380580.2 | NP_001368556.1 | 8 - 9 | 2011 | 63 | ||
| NM_001380581.2 | NP_001368557.1 | 8 - 9 | 2015 | 63 | ||
| GenBank mRNA | AF179426.1 | - | 9 - 10 | 2093 | 63 | |
More Information
Set Membership: |
|

egl-8
Genomic Map