Assay Details
| Gene Symbol: bicd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306805.2 | NP_001293734.1 | 3 - 4 | 824 | 138 | |
| NM_001306806.4 | NP_001293735.1 | 3 - 4 | 826 | 138 | ||
More Information
Set Membership: |
|

bicd-1
Genomic Map