Assay Details
| Gene Symbol: bicd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306805.2 | NP_001293734.1 | 6 - 7 | 1294 | 78 | |
| NM_001306806.4 | NP_001293735.1 | 6 - 7 | 1296 | 78 | ||
More Information
Set Membership: |
|

bicd-1
Genomic Map