Assay Details
| Gene Symbol: cest-5.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068620.6 | NP_501021.3 | 3 - 4 | 515 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cest-5.2| Gene Symbol: cest-5.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068620.6 | NP_501021.3 | 3 - 4 | 515 | 82 | |
Set Membership: |
|