Assay Details
Gene Symbol: cest-5.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068620.6 | NP_501021.3 | 5 - 6 | 761 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: cest-5.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068620.6 | NP_501021.3 | 5 - 6 | 761 | 120 |
Set Membership: |
![]() ![]() ![]() ![]() |