Assay Details
| Gene Symbol: smg-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068632.5 | NP_501033.1 | 4 - 5 | 650 | 140 | |
| GenBank mRNA | AF089729.1 | - | 4 - 5 | 670 | 140 | |
More Information
Set Membership: |
|

smg-7
Genomic Map