Assay Details
| Gene Symbol: smg-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068632.5 | NP_501033.1 | 6 - 7 | 825 | 85 | |
| GenBank mRNA | AF089729.1 | - | 6 - 7 | 845 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

smg-7| Gene Symbol: smg-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068632.5 | NP_501033.1 | 6 - 7 | 825 | 85 | |
| GenBank mRNA | AF089729.1 | - | 6 - 7 | 845 | 85 | |
Set Membership: |
|