Assay Details
| Gene Symbol: lip-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306778.3 | NP_001293707.1 | 2 - 3 | 143 | 75 | |
| NM_068652.3 | NP_501053.2 | 2 - 3 | 137 | 75 | ||
| NR_131553.1 | 2 - 3 | 137 | 75 | |||
More Information
Set Membership: |
|

lip-1
Genomic Map