Assay Details
| Gene Symbol: T22D1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068668.2 | NP_501069.2 | 5 - 6 | 985 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

T22D1.1| Gene Symbol: T22D1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068668.2 | NP_501069.2 | 5 - 6 | 985 | 71 | |
Set Membership: |
|