Assay Details
| Gene Symbol: trxr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307310.3 | NP_001294239.1 | 1 - 2 | 138 | 55 | |
| GenBank mRNA | AF148217.1 | - | 1 - 2 | 89 | 55 | |
More Information
Set Membership: |
|

trxr-1
Genomic Map