Assay Details
| Gene Symbol: trxr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306832.3 | NP_001293761.1 | 3 - 4 | 1030 | 111 | |
| NM_001307310.3 | NP_001294239.1 | 4 - 5 | 1205 | 111 | ||
| GenBank mRNA | AF148217.1 | - | 4 - 5 | 1156 | 111 | |
More Information
Set Membership: |
|

trxr-1
Genomic Map