Assay Details
| Gene Symbol: kin-33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380587.2 | NP_001367716.1 | 1 - 2 | 483 | 78 | |
| NR_151557.1 | 3 - 4 | 1411 | 78 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

kin-33| Gene Symbol: kin-33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380587.2 | NP_001367716.1 | 1 - 2 | 483 | 78 | |
| NR_151557.1 | 3 - 4 | 1411 | 78 | |||
Set Membership: |
|