Assay Details
| Gene Symbol: mltn-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269056.1 | NP_001255985.1 | 7 - 8 | 1378 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mltn-6| Gene Symbol: mltn-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269056.1 | NP_001255985.1 | 7 - 8 | 1378 | 65 | |
Set Membership: |
|