Assay Details
| Gene Symbol: dod-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313367.3 | NP_001300296.1 | 2 - 3 | 385 | 157 | |
| NM_070792.7 | NP_503193.1 | 5 - 6 | 875 | 157 | ||
More Information
Set Membership: |
|

dod-19
Genomic Map