Assay Details
| Gene Symbol: dod-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070795.4 | NP_503196.2 | 5 - 6 | 1104 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dod-20| Gene Symbol: dod-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070795.4 | NP_503196.2 | 5 - 6 | 1104 | 87 | |
Set Membership: |
|