Assay Details
| Gene Symbol: C14C6.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070840.3 | NP_503241.2 | 1 - 2 | 153 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C14C6.6| Gene Symbol: C14C6.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070840.3 | NP_503241.2 | 1 - 2 | 153 | 63 | |
Set Membership: |
|