Assay Details
| Gene Symbol: C14C6.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070840.3 | NP_503241.2 | 2 - 3 | 333 | 154 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C14C6.6| Gene Symbol: C14C6.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070840.3 | NP_503241.2 | 2 - 3 | 333 | 154 | |
Set Membership: |
|