Assay Details
| Gene Symbol: sri-73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070846.4 | NP_503247.2 | 1 - 2 | 162 | 165 | |
| NR_132479.1 | 1 - 2 | 162 | 165 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sri-73| Gene Symbol: sri-73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070846.4 | NP_503247.2 | 1 - 2 | 162 | 165 | |
| NR_132479.1 | 1 - 2 | 162 | 165 | |||
Set Membership: |
|