Assay Details
| Gene Symbol: pals-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269057.4 | NP_001255986.1 | 1 - 2 | 275 | 105 | |
| NR_132480.1 | 1 - 2 | 269 | 105 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pals-31| Gene Symbol: pals-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269057.4 | NP_001255986.1 | 1 - 2 | 275 | 105 | |
| NR_132480.1 | 1 - 2 | 269 | 105 | |||
Set Membership: |
|