Assay Details
| Gene Symbol: sri-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313122.3 | NP_001300051.1 | 2 - 3 | 203 | 78 | |
| NM_070912.4 | NP_503313.1 | 6 - 7 | 845 | 78 | ||
More Information
Set Membership: |
|

sri-4
Genomic Map