Assay Details
| Gene Symbol: srp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070914.7 | NP_503315.1 | 2 - 3 | 549 | 95 | |
| GenBank mRNA | AY525078.1 | - | 2 - 3 | 494 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srp-1| Gene Symbol: srp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070914.7 | NP_503315.1 | 2 - 3 | 549 | 95 | |
| GenBank mRNA | AY525078.1 | - | 2 - 3 | 494 | 95 | |
Set Membership: |
|