Assay Details
| Gene Symbol: srp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070914.7 | NP_503315.1 | 4 - 5 | 1016 | 95 | |
| GenBank mRNA | AY525078.1 | - | 4 - 5 | 961 | 95 | |
More Information
Set Membership: |
|

srp-1
Genomic Map