Assay Details
| Gene Symbol: flp-34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383273.1 | 10 - 11 | 3554 | 81 | ||
| NM_070964.6 | NP_503365.1 | 2 - 3 | 239 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

flp-34| Gene Symbol: flp-34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383273.1 | 10 - 11 | 3554 | 81 | ||
| NM_070964.6 | NP_503365.1 | 2 - 3 | 239 | 81 | ||
Set Membership: |
|