Assay Details
| Gene Symbol: cpr-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070982.7 | NP_503383.1 | 2 - 3 | 782 | 74 | |
| GenBank mRNA | L39896.1 | - | 2 - 3 | 729 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpr-5| Gene Symbol: cpr-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070982.7 | NP_503383.1 | 2 - 3 | 782 | 74 | |
| GenBank mRNA | L39896.1 | - | 2 - 3 | 729 | 74 | |
Set Membership: |
|