Assay Details
| Gene Symbol: cpr-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070983.6 | NP_503384.1 | 4 - 5 | 746 | 112 | |
| NR_132498.1 | 1 - 2 | 242 | 112 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpr-8| Gene Symbol: cpr-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070983.6 | NP_503384.1 | 4 - 5 | 746 | 112 | |
| NR_132498.1 | 1 - 2 | 242 | 112 | |||
Set Membership: |
|