Assay Details
| Gene Symbol: ptr-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313272.3 | NP_001300201.1 | 3 | 1745 | 84 | |
| NM_001313273.5 | NP_001300202.1 | 6 | 2024 | 84 | ||
More Information
Set Membership: |
|

ptr-16
Genomic Map