Assay Details
| Gene Symbol: Y75B7B.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070999.5 | NP_503400.1 | 3 - 4 | 296 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Y75B7B.2| Gene Symbol: Y75B7B.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_070999.5 | NP_503400.1 | 3 - 4 | 296 | 94 | |
Set Membership: |
|