Assay Details
| Gene Symbol: chil-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313242.3 | NP_001300171.1 | 3 - 4 | 610 | 111 | |
| NM_071019.5 | NP_503420.3 | 3 - 4 | 610 | 111 | ||
| Gene Symbol: hacd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071020.9 | NP_503421.1 | 4 - 5 | 642 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_071020.9 transcripts.
Set Membership: |
|

chil-14
Genomic Map