Assay Details
| Gene Symbol: srp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071127.5 | NP_503528.2 | 1 - 2 | 348 | 111 | |
| GenBank mRNA | AY525080.1 | - | 1 - 2 | 294 | 111 | |
More Information
Set Membership: |
|

srp-3
Genomic Map