Assay Details
| Gene Symbol: srp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071127.5 | NP_503528.2 | 3 - 4 | 995 | 57 | |
| GenBank mRNA | AY525080.1 | - | 3 - 4 | 941 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srp-3| Gene Symbol: srp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071127.5 | NP_503528.2 | 3 - 4 | 995 | 57 | |
| GenBank mRNA | AY525080.1 | - | 3 - 4 | 941 | 57 | |
Set Membership: |
|