Assay Details
| Gene Symbol: ckb-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071172.6 | NP_503573.1 | 2 - 3 | 284 | 70 | |
| NR_132528.1 | 1 - 2 | 132 | 70 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ckb-4| Gene Symbol: ckb-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071172.6 | NP_503573.1 | 2 - 3 | 284 | 70 | |
| NR_132528.1 | 1 - 2 | 132 | 70 | |||
Set Membership: |
|