Assay Details
| Gene Symbol: F22F7.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071173.5 | NP_503574.3 | 2 - 3 | 456 | 135 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F22F7.3| Gene Symbol: F22F7.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071173.5 | NP_503574.3 | 2 - 3 | 456 | 135 | |
Set Membership: |
|