Assay Details
| Gene Symbol: C31B8.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071388.2 | NP_503789.1 | 6 - 7 | 1006 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C31B8.7| Gene Symbol: C31B8.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071388.2 | NP_503789.1 | 6 - 7 | 1006 | 88 | |
Set Membership: |
|