Assay Details
| Gene Symbol: apx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373012.2 | NP_001360546.1 | 1 - 2 | 308 | 79 | |
| NM_071481.5 | NP_503882.2 | 1 - 2 | 315 | 79 | ||
| GenBank mRNA | U07628.1 | - | 1 - 2 | 313 | 79 | |
More Information
Set Membership: |
|

apx-1
Genomic Map