Assay Details
| Gene Symbol: apx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373012.2 | NP_001360546.1 | 4 - 5 | 753 | 95 | |
| NM_071481.5 | NP_503882.2 | 4 - 5 | 760 | 95 | ||
| NR_134544.1 | 3 - 4 | 855 | 95 | |||
| NR_134545.1 | 3 - 4 | 444 | 95 | |||
| NR_134546.1 | 1 - 2 | 198 | 95 | |||
| GenBank mRNA | U07628.1 | - | 4 - 5 | 758 | 95 | |
More Information
Set Membership: |
|

apx-1
Genomic Map