Assay Details
| Gene Symbol: apx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318263.3 | NP_001305192.1 | 1 - 2 | 283 | 66 | |
| NM_071481.5 | NP_503882.2 | 6 - 7 | 1229 | 66 | ||
| NR_134544.1 | 5 - 6 | 1324 | 66 | |||
| NR_134545.1 | 5 - 6 | 913 | 66 | |||
| NR_134546.1 | 3 - 4 | 667 | 66 | |||
| GenBank mRNA | U07628.1 | - | 6 - 7 | 1227 | 66 | |
More Information
Set Membership: |
|

apx-1
Genomic Map