Assay Details
| Gene Symbol: K08D9.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356940.3 | NP_001343687.1 | 7 - 8 | 1285 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

K08D9.2| Gene Symbol: K08D9.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356940.3 | NP_001343687.1 | 7 - 8 | 1285 | 63 | |
Set Membership: |
|