Assay Details
| Gene Symbol: slc-28.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322518.3 | NP_001309639.1 | 8 | 1500 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

slc-28.2| Gene Symbol: slc-28.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322518.3 | NP_001309639.1 | 8 | 1500 | 80 | |
Set Membership: |
|