Assay Details
| Gene Symbol: pals-33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071625.3 | NP_504026.3 | 10 - 11 | 944 | 141 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pals-33| Gene Symbol: pals-33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071625.3 | NP_504026.3 | 10 - 11 | 944 | 141 | |
Set Membership: |
|