Assay Details
| Gene Symbol: mtm-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071637.6 | NP_504038.1 | 1 - 2 | 349 | 96 | |
| GenBank mRNA | AY313178.1 | - | 1 - 2 | 347 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mtm-9| Gene Symbol: mtm-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071637.6 | NP_504038.1 | 1 - 2 | 349 | 96 | |
| GenBank mRNA | AY313178.1 | - | 1 - 2 | 347 | 96 | |
Set Membership: |
|