Assay Details
| Gene Symbol: mtm-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071637.6 | NP_504038.1 | 4 - 5 | 1516 | 69 | |
| NM_182367.6 | NP_872167.2 | 4 - 5 | 1372 | 69 | ||
| GenBank mRNA | AY313178.1 | - | 4 - 5 | 1514 | 69 | |
More Information
Set Membership: |
|

mtm-9
Genomic Map