Assay Details
| Gene Symbol: pid-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330835.3 | NP_001317846.1 | 4 - 5 | 896 | 75 | |
| NM_071761.6 | NP_504162.2 | 5 - 6 | 1112 | 75 | ||
| NR_138522.1 | 5 - 6 | 1107 | 75 | |||
More Information
Set Membership: |
|

pid-5
Genomic Map