Assay Details
| Gene Symbol: nlp-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071767.5 | NP_504168.1 | 1 - 2 | 85 | 65 | |
| NM_071768.6 | NP_504169.1 | 1 - 2 | 86 | 65 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length less than or equal to 70  Recommended Assay  Caenorhabditis elegans  Probe spans exons | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 nlp-9
nlp-9 Genomic Map
Genomic Map Back To Top
 Back To Top