Assay Details
| Gene Symbol: grl-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001330858.4 | NP_001317808.1 | 3 | 430 | 100 | |
| NM_071800.3 | NP_504201.2 | 3 - 4 | 463 | 100 | ||
More Information
Set Membership: |
|

grl-13
Genomic Map