Assay Details
| Gene Symbol: taf-10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071860.6 | NP_504261.1 | 1 - 2 | 81 | 68 | |
| Gene Symbol: K03B4.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380636.2 | NP_001367844.1 | 1 - 2 | 91 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380636.2 transcripts.
Set Membership: |
|

taf-10
Genomic Map