Assay Details
| Gene Symbol: F39G3.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071872.5 | NP_504273.2 | 4 - 5 | 709 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F39G3.2| Gene Symbol: F39G3.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071872.5 | NP_504273.2 | 4 - 5 | 709 | 71 | |
Set Membership: |
|