Assay Details
| Gene Symbol: tam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071934.6 | NP_504335.1 | 2 - 3 | 1427 | 65 | |
| GenBank mRNA | AF164625.1 | - | 1 - 2 | 41 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tam-1| Gene Symbol: tam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071934.6 | NP_504335.1 | 2 - 3 | 1427 | 65 | |
| GenBank mRNA | AF164625.1 | - | 1 - 2 | 41 | 65 | |
Set Membership: |
|