Assay Details
| Gene Symbol: tam-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071934.6 | NP_504335.1 | 3 - 4 | 1533 | 68 | |
| GenBank mRNA | AF164625.1 | - | 2 - 3 | 147 | 68 | |
More Information
Set Membership: |
|

tam-1
Genomic Map